Learn More
Medchemexpress LLC Thalidomide-O-C8-Boc | 2225148-52-5 | MFCD32701934 | 95.1% | 486.56 g·mol⁻¹ | C26H34N2O7 | 5 MG
SDP
Catalog No. 5000262797
Encompass_Preferred
Thalidomide-O-C8-Boc is a thalidomide-derived Cereblon (CRBN) ligand for targeted protein degradation research. It is designed to be connected to a protein-targeting ligand via a linker to form PROTAC molecules and is provided as a research-grade chemical with manufacturer-reported purity (~95.1%).
  • Acts as a cereblon ligand to recruit CRBN.
  • Designed for conjugation via a linker to form PROTACs.
  • Supplied at research-grade purity (~95.1%).
  • Molecular formula C26H34N2O7; molecular weight 486.56 g·mol⁻¹.
  • Available in milligram-scale quantities suitable for synthesis and screening.
Catalog No. 50-002-62797 Supplier Medchemexpress LLC Supplier No. HY1311595MG
Thalidomide-O-C8-Boc is a thalidomide-derived Cereblon (CRBN) ligand for targeted protein degradation research. It is designed to be connected to a protein-targeting ligand via a linker to form PROTAC molecules and is provided as a research-grade chemical with manufacturer-reported purity (~95.1%).
  • Acts as a cereblon ligand to recruit CRBN.
  • Designed for conjugation via a linker to form PROTACs.
  • Supplied at research-grade purity (~95.1%).
  • Molecular formula C26H34N2O7; molecular weight 486.56 g·mol⁻¹.
  • Available in milligram-scale quantities suitable for synthesis and screening.