Learn More
Medchemexpress LLC Thalidomide-O-C8-Boc | 2225148-52-5 | MFCD32701934 | 95.1% | C26H34N2O7 | 10MG
SDP
Catalog No. 5000261825
Encompass_Preferred
Thalidomide-O-C8-Boc is a thalidomide-derived Cereblon (CRBN) ligand intended for use as the E3 ligase-recruiting moiety in PROTAC design. It features an O-linked eight-carbon spacer with a Boc-protected terminus to enable linker attachment. Manufacturer data report molecular formula C26H34N2O7, molecular weight 486.56, and typical purity ~95%.
  • Provides an O-linked C8 spacer with a Boc-protected terminus for linker attachment.
  • Suitable for recruiting Cereblon (CRBN) as the E3 ligase in degrader strategies.
  • Reported purity approximately 95.1%.
  • High solubility in DMSO (100 mg/mL); manufacturer lists example in vivo formulations.
  • Recommended storage: solid at -20°C under nitrogen; in solution -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-61825 Supplier Medchemexpress LLC Supplier No. HY13115910MG
Thalidomide-O-C8-Boc is a thalidomide-derived Cereblon (CRBN) ligand intended for use as the E3 ligase-recruiting moiety in PROTAC design. It features an O-linked eight-carbon spacer with a Boc-protected terminus to enable linker attachment. Manufacturer data report molecular formula C26H34N2O7, molecular weight 486.56, and typical purity ~95%.
  • Provides an O-linked C8 spacer with a Boc-protected terminus for linker attachment.
  • Suitable for recruiting Cereblon (CRBN) as the E3 ligase in degrader strategies.
  • Reported purity approximately 95.1%.
  • High solubility in DMSO (100 mg/mL); manufacturer lists example in vivo formulations.
  • Recommended storage: solid at -20°C under nitrogen; in solution -80°C (6 months) or -20°C (1 month).