Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-C8-COOH | 2225148-51-4 | 97.4% | 430.45 g/mol | C22H26N2O7 | 5 MG
SDP

Catalog No. 5000263006
Encompass_Preferred

Thalidomide-O-C8-COOH is a thalidomide-derived cereblon ligand with a carboxy-terminated eight-carbon linker, developed for use as an E3 ligase recruiter in targeted protein degradation. It is provided as a high-purity solid optimized for conjugation into PROTACs and other bifunctional degraders, with documented solubility and storage recommendations for research workflows.

  • High purity (97.4%) suitable for research applications.
  • Carboxy-terminated C8 linker for convenient coupling chemistry.
  • Molecular weight 430.45 g/mol; formula C22H26N2O7.
  • High solubility in DMSO for preparation of concentrated stock solutions.
  • Storage and solvent stability data provided for reliable handling.

Catalog No. 50-002-63006 Supplier Medchemexpress LLC Supplier No. HY1309525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-C8-COOH is a thalidomide-derived cereblon ligand with a carboxy-terminated eight-carbon linker, developed for use as an E3 ligase recruiter in targeted protein degradation. It is provided as a high-purity solid optimized for conjugation into PROTACs and other bifunctional degraders, with documented solubility and storage recommendations for research workflows.

  • High purity (97.4%) suitable for research applications.
  • Carboxy-terminated C8 linker for convenient coupling chemistry.
  • Molecular weight 430.45 g/mol; formula C22H26N2O7.
  • High solubility in DMSO for preparation of concentrated stock solutions.
  • Storage and solvent stability data provided for reliable handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.