Learn More
Medchemexpress LLC Thalidomide-O-C8-COOH | 2225148-51-4 | 97.4% | C22H26N2O7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13095210MG
Encompass_Preferred
Thalidomide-O-C8-COOH is a thalidomide-derived cereblon (CRBN) ligand supplied as a solid research reagent for use in PROTAC design and chemical biology studies. It has molecular formula C22H26N2O7, molecular weight 430.45 g/mol, and a reported purity of 97.44%.
  • Used as a cereblon E3 ligase recruiting moiety in PROTAC synthesis.
  • White to off-white solid suitable for laboratory handling.
  • Reported purity of 97.4% for research applications.
  • Storage guidance provided for long-term and in-solvent conditions.
  • Available in small research quantities for assay development.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-61890