Learn More
Selleck Chemical LLC Thalidomide-O-COOH
SDP
Supplier:  Selleck Chemical LLC S679425MG
Encompass_Preferred
Thalidomide-O-COOH (Cereblon ligand 3 E3 ligase Ligand 3) a Thalidomide-based Cereblon (CRBN) ligand is used in the recruitment of CRBN protein Thalidomide-O-COOH (Cereblon ligand 3) can be connected to the ligand for protein by a linker to form PROTACs (Proteolysis Targeting Chimera)
Catalog No. 50-000-02560