Learn More
Medchemexpress LLC Thalidomide-O-COOH | 1061605-21-7 | MFCD31560477 | >98.0% | 332.27 | C15H12N2O7 | 10 G
SDP
Catalog No. 5000245033
Encompass_Preferred
Thalidomide-O-COOH is a thalidomide-derived cereblon (CRBN) ligand used as an E3 ligase recruiting moiety in PROTAC and targeted protein degradation design. The compound is supplied as a solid with molecular formula C15H12N2O7 and molecular weight 332.27 g/mol (CAS 1061605-21-7).
  • Serves as a cereblon (CRBN) binding ligand for PROTAC construction.
  • Suitable for medicinal chemistry and chemical biology research.
  • Provided in multiple laboratory-scale pack sizes for synthesis and screening.
  • Characterized by manufacturer datasheet and SDS for handling and storage guidance.
Catalog No. 50-002-45033 Supplier Medchemexpress LLC Supplier No. HY10359710G
Thalidomide-O-COOH is a thalidomide-derived cereblon (CRBN) ligand used as an E3 ligase recruiting moiety in PROTAC and targeted protein degradation design. The compound is supplied as a solid with molecular formula C15H12N2O7 and molecular weight 332.27 g/mol (CAS 1061605-21-7).
  • Serves as a cereblon (CRBN) binding ligand for PROTAC construction.
  • Suitable for medicinal chemistry and chemical biology research.
  • Provided in multiple laboratory-scale pack sizes for synthesis and screening.
  • Characterized by manufacturer datasheet and SDS for handling and storage guidance.