Learn More
Medchemexpress LLC Thalidomide-O-COOH | 1061605-21-7 | MFCD31560477 | 99.9% | 332.26 g/mol | C15H12N2O7 | 5 G
SDP
Catalog No. 5000244618
Encompass_Preferred
Thalidomide-O-COOH is a thalidomide-derived carboxylic acid used as a cereblon (CRBN) ligand building block in PROTAC design and E3 ligase recruitment studies. It is supplied as a high-purity research chemical in small laboratory pack sizes for medicinal chemistry and chemical biology applications.
  • Thalidomide-derived cereblon ligand for PROTAC design.
  • Carboxylic acid functionality enables linker attachment and derivatization.
  • Supplied as a solid suitable for standard laboratory storage.
  • Provided in small research pack sizes for synthesis and assay development.
  • High reported purity supports use in chemical biology experiments.
Catalog No. 50-002-44618 Supplier Medchemexpress LLC Supplier No. HY1035975G
Thalidomide-O-COOH is a thalidomide-derived carboxylic acid used as a cereblon (CRBN) ligand building block in PROTAC design and E3 ligase recruitment studies. It is supplied as a high-purity research chemical in small laboratory pack sizes for medicinal chemistry and chemical biology applications.
  • Thalidomide-derived cereblon ligand for PROTAC design.
  • Carboxylic acid functionality enables linker attachment and derivatization.
  • Supplied as a solid suitable for standard laboratory storage.
  • Provided in small research pack sizes for synthesis and assay development.
  • High reported purity supports use in chemical biology experiments.