Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-COOH | 1061605-21-7 | MFCD31560477 | 99.9% | 332.27 | C15H12N2O7 | 25 G
SDP

Catalog No. 5000245004
Encompass_Preferred

Thalidomide-O-COOH is a thalidomide-derived cereblon (CRBN) ligand used as an E3 ligase recruiter in PROTAC and chemical biology research. It is supplied as a solid research reagent with high purity and characterized molecular properties for use in biochemical and cellular assays.

  • High purity: 99.85%.
  • Molecular formula: C15H12N2O7.
  • Molecular weight: 332.27 g/mol.
  • Appearance: solid, white to off-white.
  • Available package sizes include 25 G for bulk research use.
  • Intended use: cereblon recruitment and PROTAC design in research applications.

Catalog No. 50-002-45004 Supplier Medchemexpress LLC Supplier No. HY10359725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-O-COOH is a thalidomide-derived cereblon (CRBN) ligand used as an E3 ligase recruiter in PROTAC and chemical biology research. It is supplied as a solid research reagent with high purity and characterized molecular properties for use in biochemical and cellular assays.

  • High purity: 99.85%.
  • Molecular formula: C15H12N2O7.
  • Molecular weight: 332.27 g/mol.
  • Appearance: solid, white to off-white.
  • Available package sizes include 25 G for bulk research use.
  • Intended use: cereblon recruitment and PROTAC design in research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.