Learn More
Selleck Chemical LLC Thalidomide-O-COOH
SDP
Catalog No. 5000001770
Encompass_Preferred
Thalidomide-O-COOH (Cereblon ligand 3 E3 ligase Ligand 3) a Thalidomide-based Cereblon (CRBN) ligand is used in the recruitment of CRBN protein Thalidomide-O-COOH (Cereblon ligand 3) can be connected to the ligand for protein by a linker to form PROTACs (Proteolysis Targeting Chimera)
Catalog No. 50-000-01770 Supplier Selleck Chemical LLC Supplier No. S67945MG
Thalidomide-O-COOH (Cereblon ligand 3 E3 ligase Ligand 3) a Thalidomide-based Cereblon (CRBN) ligand is used in the recruitment of CRBN protein Thalidomide-O-COOH (Cereblon ligand 3) can be connected to the ligand for protein by a linker to form PROTACs (Proteolysis Targeting Chimera)