Learn More
Medchemexpress LLC Thalidomide-O-PEG2-propargyl | 2098487-52-4 | 98.3% | 400.38 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY126458100MG
Encompass_Preferred
Thalidomide-O-PEG2-propargyl is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 2-unit PEG linker used in PROTAC technology. It is also a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Contains 2-unit PEG linker
  • Used in PROTAC technology
  • Click chemistry reagent with an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-004-36931