Learn More
Medchemexpress LLC Thalidomide-O-PEG2-propargyl | 2098487-52-4 | 98.3% | 400.38 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY126458500MG
Encompass_Preferred
Thalidomide-O-PEG2-propargyl, also known as E3 ligase Ligand-Linker Conjugates 32, is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a 2-unit PEG linker, making it suitable for PROTAC technology. This product also functions as a click chemistry reagent, containing an Alkyne group and capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Used in PROTAC technology.
  • Click chemistry reagent with an alkyne group.
  • Undergoes CuAAc with azide groups.
Catalog No. 50-004-38066