Learn More
Medchemexpress LLC Thalidomide-O-PEG4-azide | 2380318-57-8 | 98.0% | C23H29N5O9 | 100 MG
SDP
Catalog No. 5000354994
Encompass_Preferred
Thalidomide-O-PEG4-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates the thalidomide based cereblon ligand and a linker used in PROTAC technology. It is a click chemistry reagent, containing an azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Used in PROTAC technology
  • Click chemistry reagent containing an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Molecular weight of 519.50
  • Appears as a colorless to light yellow oil
Catalog No. 50-003-54994 Supplier Medchemexpress LLC Supplier No. HY140844100MG
Thalidomide-O-PEG4-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates the thalidomide based cereblon ligand and a linker used in PROTAC technology. It is a click chemistry reagent, containing an azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Used in PROTAC technology
  • Click chemistry reagent containing an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Molecular weight of 519.50
  • Appears as a colorless to light yellow oil