Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-O-PEG4-azide | 2380318-57-8 | 98.0% | C23H29N5O9 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY140844

Encompass_Preferred

Thalidomide-O-PEG4-azide is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. It is a click chemistry reagent containing an Azide group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. This product is for research use only.

  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Contains an Azide group for click chemistry
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Target: Cereblon

Catalog No. 50-003-52517


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.