Learn More
Medchemexpress LLC Thalidomide-O-PEG4-azide | 2380318-57-8 | 98.0% | C23H29N5O9 | 50 MG
SDP
Catalog No. 5000355108
Encompass_Preferred
Thalidomide-O-PEG4-azide is a synthesized E3 ligase ligand-linker conjugate that integrates a Thalidomide-based cereblon ligand with a linker used in PROTAC technology. This compound is intended for research use only.
  • Acts as a click chemistry reagent due to its Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups
Catalog No. 50-003-55108 Supplier Medchemexpress LLC Supplier No. HY14084450MG
Thalidomide-O-PEG4-azide is a synthesized E3 ligase ligand-linker conjugate that integrates a Thalidomide-based cereblon ligand with a linker used in PROTAC technology. This compound is intended for research use only.
  • Acts as a click chemistry reagent due to its Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups