Learn More
Medchemexpress LLC Thalidomide-O-PEG4-Boc | 2411681-87-1 | C28H38N2O11 | 100 MG
SDP
Catalog No. 5000353381
Encompass_Preferred
Thalidomide-O-PEG4-Boc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide-based cereblon ligand and a linker used in PROTAC technology. This molecule is designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Targets E3 Ligase Ligand-Linker Conjugates, Apoptosis, and Autophagy
  • Targets Cereblon with an IC50 & Target affinity
  • Molecular weight of 578.61
Catalog No. 50-003-53381 Supplier Medchemexpress LLC Supplier No. HY141014100MG
Thalidomide-O-PEG4-Boc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide-based cereblon ligand and a linker used in PROTAC technology. This molecule is designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Used in PROTAC technology
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Targets E3 Ligase Ligand-Linker Conjugates, Apoptosis, and Autophagy
  • Targets Cereblon with an IC50 & Target affinity
  • Molecular weight of 578.61