Learn More
Medchemexpress LLC Thalidomide-O-PEG4-Boc | 2411681-87-1 | C28H38N2O11 | 25MG
SDP
Catalog No. 5000352902
Encompass_Preferred
Thalidomide-O-PEG4-Boc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
  • Incorporates thalidomide-based cereblon ligand.
  • Utilizes a linker in PROTAC technology.
  • Contains two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase, and the other is for the target protein.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in PROTAC technology.
Catalog No. 50-003-52902 Supplier Medchemexpress LLC Supplier No. 25MGHY141014
Thalidomide-O-PEG4-Boc is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
  • Incorporates thalidomide-based cereblon ligand.
  • Utilizes a linker in PROTAC technology.
  • Contains two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase, and the other is for the target protein.
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in PROTAC technology.