Learn More
Medchemexpress LLC Thalidomide-PEG2-C2-NH2 hydrochloride | 2245697-87-2 | 99.9% | 10 MG
SDP
Catalog No. 5000341117
Encompass_Preferred
Thalidomide-PEG2-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates a thalidomide based cereblon ligand and a 2-unit PEG linker, making it suitable for PROTAC technology applications. This product is for research use only.
  • Used in PROTAC technology
  • Suitable for autophagy research
  • Applicable for apoptosis studies
  • E3 ligase ligand-linker conjugate
  • Targets cereblon
  • Relevant for cancer targeted therapy research
  • Used in cancer metabolism and metastasis studies
  • Acts as an inhibitor
Catalog No. 50-003-41117 Supplier Medchemexpress LLC Supplier No. HY129703B10MG
Thalidomide-PEG2-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates a thalidomide based cereblon ligand and a 2-unit PEG linker, making it suitable for PROTAC technology applications. This product is for research use only.
  • Used in PROTAC technology
  • Suitable for autophagy research
  • Applicable for apoptosis studies
  • E3 ligase ligand-linker conjugate
  • Targets cereblon
  • Relevant for cancer targeted therapy research
  • Used in cancer metabolism and metastasis studies
  • Acts as an inhibitor