Learn More
Medchemexpress LLC Thalidomide-PEG2-C2-NH2 (hydrochloride) | 2245697-87-2 | >99.9% | 100 MG
SDP
Catalog No. 5000341116
Encompass_Preferred
Thalidomide-PEG2-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a 2-unit PEG linker used in PROTAC technology. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in PROTAC technology.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
  • Incorporates thalidomide based cereblon ligand.
  • Features a 2-unit PEG linker.
  • For research use only.
Catalog No. 50-003-41116 Supplier Medchemexpress LLC Supplier No. HY129703B100MG
Thalidomide-PEG2-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a 2-unit PEG linker used in PROTAC technology. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in PROTAC technology.
  • Exploits the intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
  • Incorporates thalidomide based cereblon ligand.
  • Features a 2-unit PEG linker.
  • For research use only.