Learn More
Medchemexpress LLC Thalidomide-PEG2-C2-NH2 hydrochloride | 2245697-87-2 | 99.9% | 25 MG
SDP
Catalog No. 5000340797
Encompass_Preferred
Thalidomide-PEG2-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a 2-unit PEG linker, making it suitable for use in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a cereblon ligand
  • Features a 2-unit PEG linker
  • Utilized in PROTAC technology
  • For research use only
Catalog No. 50-003-40797 Supplier Medchemexpress LLC Supplier No. 25MGHY129703B
Thalidomide-PEG2-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a 2-unit PEG linker, making it suitable for use in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a cereblon ligand
  • Features a 2-unit PEG linker
  • Utilized in PROTAC technology
  • For research use only