Learn More
Medchemexpress LLC Thalidomide-PEG2-C2-NH2 hydrochloride | 2245697-87-2 | 99.9% | 50 MG
SDP
Catalog No. 5000340363
Encompass_Preferred
Thalidomide-PEG2-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 2-unit PEG linker used in PROTAC technology. It is for research use only and not sold to patients.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Incorporates Thalidomide based cereblon ligand.
  • Features a 2-unit PEG linker.
  • Utilized in PROTAC technology.
  • Exploits intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
  • Targets Cereblon.
  • For research use only.
Catalog No. 50-003-40363 Supplier Medchemexpress LLC Supplier No. HY129703B50MG
Thalidomide-PEG2-C2-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and 2-unit PEG linker used in PROTAC technology. It is for research use only and not sold to patients.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Incorporates Thalidomide based cereblon ligand.
  • Features a 2-unit PEG linker.
  • Utilized in PROTAC technology.
  • Exploits intracellular ubiquitin-proteasome system.
  • Selectively degrades target proteins.
  • Targets Cereblon.
  • For research use only.