Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-PEG2-C2-NH2 (TFA) | 2097509-36-7 | 99.1% | 100 MG
SDP

Catalog No. 5000338522
Encompass_Preferred

Thalidomide-PEG2-C2-NH2 (TFA) is a high-purity chemical compound, identified as an E3 ligase ligand-linker conjugate. It is designed for laboratory research purposes and exhibits chemical stability under recommended storage conditions.

  • High purity of 99.1% (HPLC)
  • Stable under recommended storage conditions
  • E3 ligase ligand-linker conjugate
  • Suitable for laboratory research
  • Recommended storage at -20°C under nitrogen
  • For solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Catalog No. 50-003-38522 Supplier Medchemexpress LLC Supplier No. HY129703A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-PEG2-C2-NH2 (TFA) is a high-purity chemical compound, identified as an E3 ligase ligand-linker conjugate. It is designed for laboratory research purposes and exhibits chemical stability under recommended storage conditions.

  • High purity of 99.1% (HPLC)
  • Stable under recommended storage conditions
  • E3 ligase ligand-linker conjugate
  • Suitable for laboratory research
  • Recommended storage at -20°C under nitrogen
  • For solutions, store at -80°C for 6 months or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.