Learn More
Medchemexpress LLC Thalidomide-PEG3-COOH | 2682112-08-7 | 96.95% | 434.40 | 25 MG
SDP
Catalog No. 5000349217
Encompass_Preferred
Thalidomide-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology, which exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Used in PROTAC technology
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Targets E3 ligase ligand-linker conjugates, apoptosis, and autophagy
  • Targets Cereblon
Catalog No. 50-003-49217 Supplier Medchemexpress LLC Supplier No. 25MGHY138775
Thalidomide-PEG3-COOH is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology, which exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide based cereblon ligand
  • Used in PROTAC technology
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Targets E3 ligase ligand-linker conjugates, apoptosis, and autophagy
  • Targets Cereblon