Learn More
Medchemexpress LLC Thalidomide-PEG4-COOH | 2828438-28-2 | 95.4% | 478.45 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY138774
Encompass_Preferred
Thalidomide-PEG4-COOH is a synthesized E3 ligase ligand-linker conjugate, incorporating a Thalidomide-based cereblon ligand and a linker utilized in PROTAC technology. This product is intended for research use only.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates a Thalidomide-based cereblon ligand
  • Utilized in PROTAC technology
  • Targets cereblon
  • Associated with cancer, cancer targeted therapy, cancer metabolism, and metastasis research
  • Relevant to autophagy and apoptosis studies
  • Functions as an inhibitor
Catalog No. 50-003-48461