Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-PEG5-COOH | 2688100-28-7 | 95.9% | 522.50 | 25 MG
SDP

Catalog No. 5000348666
Encompass_Preferred

Thalidomide-PEG5-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a thalidomide-based cereblon ligand and a linker utilized in PROTAC technology. This product is intended for research purposes only and not for medical, clinical diagnostic, or food use in humans or animals.

  • Purity: 95.90%
  • Appearance: Oil
  • Color: Light yellow to brown
  • Solubility: Soluble in DMSO at 50 mg/mL (95.69 mM)

Catalog No. 50-003-48666 Supplier Medchemexpress LLC Supplier No. 25MGHY138773
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-PEG5-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates a thalidomide-based cereblon ligand and a linker utilized in PROTAC technology. This product is intended for research purposes only and not for medical, clinical diagnostic, or food use in humans or animals.

  • Purity: 95.90%
  • Appearance: Oil
  • Color: Light yellow to brown
  • Solubility: Soluble in DMSO at 50 mg/mL (95.69 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.