Learn More
Medchemexpress LLC Thalidomide-PEG5-NH2 hydrochloride | 2703775-06-6 | C23H31N3O9 | 10MG
SDP
Supplier:  Medchemexpress LLC HY138784A10MG
Encompass_Preferred
Thalidomide-PEG5-NH2 hydrochloride is a thalidomide-derived cereblon ligand-linker conjugate intended for PROTAC development and related chemical biology applications. It links a thalidomide-based CRBN-binding moiety to a PEG5 primary amine, enabling coupling to target-binding warheads while providing documentation for synthetic and screening use.
  • Thalidomide-based cereblon ligand for CRBN recruitment.
  • PEG5 amine linker enabling facile conjugation to warheads.
  • Provided as a hydrochloride salt for the amine functionality.
  • Available in small milligram quantities suitable for medicinal chemistry.
  • Supplied with purity documentation and safety data sheet.
Catalog No. 50-002-60550