Learn More
Medchemexpress LLC Thalidomide-PEG5-NH2 hydrochloride | 2703775-06-6 | MFCD34550803 | 95.2% | 529.97 g·mol⁻¹ | C23H32ClN3O9 | 5 MG
SDP
Catalog No. 5000222061
Encompass_Preferred
Thalidomide-PEG5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon (CRBN) binding moiety with a PEG5 amine linker, supplied as the hydrochloride salt. It is intended for use in PROTAC design and chemical biology research involving CRBN-mediated protein degradation.
  • Cereblon-binding thalidomide-derived ligand.
  • PEG5 amine linker provides a flexible spacer for conjugation.
  • Hydrochloride salt improves solubility and handling.
  • Characterized by CAS 2703775-06-6 and molecular weight 529.97 g·mol⁻¹.
  • Purity approximately 95.2% by LCMS.
  • Intended for research use in targeted protein degradation studies.
Catalog No. 50-002-22061 Supplier Medchemexpress LLC Supplier No. HY138784A5MG
Thalidomide-PEG5-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-derived cereblon (CRBN) binding moiety with a PEG5 amine linker, supplied as the hydrochloride salt. It is intended for use in PROTAC design and chemical biology research involving CRBN-mediated protein degradation.
  • Cereblon-binding thalidomide-derived ligand.
  • PEG5 amine linker provides a flexible spacer for conjugation.
  • Hydrochloride salt improves solubility and handling.
  • Characterized by CAS 2703775-06-6 and molecular weight 529.97 g·mol⁻¹.
  • Purity approximately 95.2% by LCMS.
  • Intended for research use in targeted protein degradation studies.