Learn More
Medchemexpress LLC Thalidomide-pinacolborane | 2229725-98-6 | 95.0% | 384.19 g·mol⁻¹ | C19H21BN2O6 | 10 MG
SDP
Catalog No. 5000282110
Encompass_Preferred
Thalidomide-pinacolborane is a thalidomide-derived small molecule bearing a pinacol boronic ester (Bpin). It serves as an E3 ligase (cereblon) ligand useful in targeted protein degradation (PROTAC) research and as a synthetic intermediate for cross-coupling chemistries.
Catalog No. 50-002-82110 Supplier Medchemexpress LLC Supplier No. HYW24428210MG
Thalidomide-pinacolborane is a thalidomide-derived small molecule bearing a pinacol boronic ester (Bpin). It serves as an E3 ligase (cereblon) ligand useful in targeted protein degradation (PROTAC) research and as a synthetic intermediate for cross-coupling chemistries.