Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-pinacolborane | 2229725-98-6 | 97.0% | 384.19 | C19H21BN2O6 | 100 MG
SDP

Catalog No. 5000282572
Encompass_Preferred

Thalidomide-pinacolborane is a thalidomide analogue that incorporates a boronic pinacol ester (Bpin). It is used as an E3 ligase ligand in PROTAC research and as a boronate building block for cross-coupling chemistry. Intended for research use only.

  • Contains a boronic pinacol ester (Bpin) suitable for Suzuki cross-coupling.
  • Acts as a ligand for E3 ligase in PROTAC applications.
  • Characterized identity with CAS 2229725-98-6 and molecular weight 384.19.
  • Purity typically reported as 97.0% by supplier.
  • Recommended storage under nitrogen at -20°C; in solvent, store at -80°C for up to 6 months.

Catalog No. 50-002-82572 Supplier Medchemexpress LLC Supplier No. HYW244282100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-pinacolborane is a thalidomide analogue that incorporates a boronic pinacol ester (Bpin). It is used as an E3 ligase ligand in PROTAC research and as a boronate building block for cross-coupling chemistry. Intended for research use only.

  • Contains a boronic pinacol ester (Bpin) suitable for Suzuki cross-coupling.
  • Acts as a ligand for E3 ligase in PROTAC applications.
  • Characterized identity with CAS 2229725-98-6 and molecular weight 384.19.
  • Purity typically reported as 97.0% by supplier.
  • Recommended storage under nitrogen at -20°C; in solvent, store at -80°C for up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.