Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-pinacolborane | 2229725-98-6 | 97.0% | 384.19 g·mol⁻1 | C19H21BN2O6 | 25 MG
SDP

Catalog No. 5000280693
Encompass_Preferred

Thalidomide-pinacolborane is a thalidomide-derived boronic pinacol ester used as a cereblon (CRBN) ligand for E3 ligase recruitment in PROTAC design and for synthetic transformations such as Suzuki cross-coupling. It is supplied as a solid research reagent intended for use in ligand design, chemical synthesis, and targeted protein degradation studies.

  • Contains boronic pinacol ester (Bpin) functionality for Suzuki coupling.
  • Functions as a cereblon E3 ligase ligand suitable for PROTAC applications.
  • Molecular formula C19H21BN2O6; molecular weight 384.19 g·mol⁻1.
  • CAS number 2229725-98-6.
  • Purity 97.0% as listed by the supplier.
  • Provided as a 25 MG research pack.

Catalog No. 50-002-80693 Supplier Medchemexpress LLC Supplier No. HYW24428225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-pinacolborane is a thalidomide-derived boronic pinacol ester used as a cereblon (CRBN) ligand for E3 ligase recruitment in PROTAC design and for synthetic transformations such as Suzuki cross-coupling. It is supplied as a solid research reagent intended for use in ligand design, chemical synthesis, and targeted protein degradation studies.

  • Contains boronic pinacol ester (Bpin) functionality for Suzuki coupling.
  • Functions as a cereblon E3 ligase ligand suitable for PROTAC applications.
  • Molecular formula C19H21BN2O6; molecular weight 384.19 g·mol⁻1.
  • CAS number 2229725-98-6.
  • Purity 97.0% as listed by the supplier.
  • Provided as a 25 MG research pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.