Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-pinacolborane | 2229725-98-6 | 97.0% | 384.19 g/mol | C19H21BN2O6 | 250 MG
SDP

Catalog No. 5000280543
Encompass_Preferred

Thalidomide-pinacolborane is a thalidomide analogue that incorporates a boronic pinacol ester (Bpin). It is used as a synthetic intermediate for cross-coupling chemistry and as an E3 ligase (cereblon) ligand in chemical biology and targeted protein degradation research.

  • Contains a boronic pinacol ester suitable for Suzuki-Miyaura cross-coupling.
  • Functions as an E3 ligase (cereblon) ligand for PROTAC and degradation studies.
  • High purity appropriate for research applications.
  • Available in research-scale quantities for assay development.
  • Molecular formula C19H21BN2O6; molecular weight 384.19 g/mol.

Catalog No. 50-002-80543 Supplier Medchemexpress LLC Supplier No. HYW244282250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-pinacolborane is a thalidomide analogue that incorporates a boronic pinacol ester (Bpin). It is used as a synthetic intermediate for cross-coupling chemistry and as an E3 ligase (cereblon) ligand in chemical biology and targeted protein degradation research.

  • Contains a boronic pinacol ester suitable for Suzuki-Miyaura cross-coupling.
  • Functions as an E3 ligase (cereblon) ligand for PROTAC and degradation studies.
  • High purity appropriate for research applications.
  • Available in research-scale quantities for assay development.
  • Molecular formula C19H21BN2O6; molecular weight 384.19 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.