Learn More
Medchemexpress LLC Thalidomide-pinacolborane | 2229725-98-6 | 97.0% | 384.19 g/mol | C19H21BN2O6 | 50 MG
SDP
Catalog No. 5000282573
Encompass_Preferred
Thalidomide-pinacolborane is a thalidomide analogue containing a pinacol boronate (Bpin) group, supplied as a research reagent for use in synthetic chemistry and chemical biology. It is suitable as a building block for cross-coupling reactions and as a probe to investigate E3 ligase-mediated ubiquitination in degradation studies.
  • Contains a pinacol boronate functional group for cross-coupling chemistry.
  • Useful as a synthetic intermediate in Suzuki-Miyaura reactions.
  • Acts as a thalidomide analogue for studying E3 ligase activity.
  • Supplied as a high-purity research chemical suitable for analytical and preparative use.
  • Provided in small-quantity packaging for laboratory experiments.
Catalog No. 50-002-82573 Supplier Medchemexpress LLC Supplier No. HYW24428250MG
Thalidomide-pinacolborane is a thalidomide analogue containing a pinacol boronate (Bpin) group, supplied as a research reagent for use in synthetic chemistry and chemical biology. It is suitable as a building block for cross-coupling reactions and as a probe to investigate E3 ligase-mediated ubiquitination in degradation studies.
  • Contains a pinacol boronate functional group for cross-coupling chemistry.
  • Useful as a synthetic intermediate in Suzuki-Miyaura reactions.
  • Acts as a thalidomide analogue for studying E3 ligase activity.
  • Supplied as a high-purity research chemical suitable for analytical and preparative use.
  • Provided in small-quantity packaging for laboratory experiments.