Learn More
Medchemexpress LLC Thalidomide-pinacolborane | 2229725-98-6 | N/A | 97.0% | 384.19 g/mol | C19H21BN2O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW2442825MG
Encompass_Preferred
Thalidomide-pinacolborane is a thalidomide analogue bearing a pinacolborane (Bpin) group, used as a synthetic intermediate and as a ligand for E3 ligase in PROTAC research. Molecular formula C19H21BN2O6, molecular weight 384.19 g/mol. Manufacturer documentation lists CAS 2229725-98-6 and purity 97.0%. Recommended storage is -20°C under nitrogen; in solvent, -80°C (up to 6 months) or -20°C (up to 1 month) under nitrogen.
  • Functionalized thalidomide analogue useful as a synthetic intermediate.
  • Serves as a ligand for E3 ligase in PROTAC applications.
  • High purity (97.0%) suitable for research use.
  • Stable when stored under nitrogen at recommended temperatures.
  • Available in small research sizes down to 5 mg.
Catalog No. 50-002-80694