Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-Piperazine 5-fluoride | 2222114-22-7 | 96.47% | 360.34 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY134983

Encompass_Preferred

Thalidomide-Piperazine 5-fluoride is a synthesized E3 ligase ligand-linker conjugate. It incorporates a cereblon ligand and a linker utilized in PROTAC technology, designed for research purposes.

  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates cereblon ligand
  • Linker utilized in PROTAC technology
  • Intended for research use only
  • Purity: 96.47%
  • Molecular weight: 360.34
  • Solid appearance
  • Color: Light yellow to green yellow
  • Soluble in DMSO

Catalog No. 50-003-66043


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.