Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-Piperazine 5-fluoride | 2222114-22-7 | >96.5% | 360.34 | 5 G
SDP

Supplier:  Medchemexpress LLC 5GHY134983

Encompass_Preferred

Thalidomide-Piperazine 5-fluoride is a synthesized E3 ligase ligand-linker conjugate. Its intended use is for research purposes, specifically in the field of PROTAC (Proteolysis Targeting Chimeras) technology. It incorporates a thalidomide-based cereblon ligand and a linker. This product is not for sale to patients.

  • Appearance: Solid, light yellow to green yellow
  • Solubility: Soluble in DMSO (5.83 mg/mL or 16.18 mM)
  • Storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month (protect from light)
  • Related research areas: Cancer, PROTAC, autophagy, apoptosis, E3 ligase ligand-linker conjugates
  • Cereblon ligand: Incorporates a thalidomide-based cereblon ligand
  • PROTAC technology: Utilizes a linker commonly used in PROTAC technology

Catalog No. 50-003-65969


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.