Learn More
Medchemexpress LLC Thalidomide-piperazine 5-fluoride hydrochloride | 2222114-23-8 | MFCD32203462 | 98.9% | 396.80 g/mol | C17H18ClFN4O4 | 1 G
SDP
Catalog No. 5000282925
Encompass_Preferred
Thalidomide-piperazine 5-fluoride hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as a hydrochloride salt for use in PROTAC synthesis and related cereblon-targeted chemical biology research. It is intended for research use and is provided with documented purity and stability data for handling and storage.
  • Thalidomide-based cereblon ligand for PROTAC synthesis.
  • Hydrochloride salt form for improved handling and solubility.
  • High purity suitable for synthetic and analytical workflows.
  • Documented stability under cold storage conditions.
  • Provided in a small-scale research package for medicinal chemistry.
Catalog No. 50-002-82925 Supplier Medchemexpress LLC Supplier No. HYW1454361G
Thalidomide-piperazine 5-fluoride hydrochloride is a thalidomide-derived cereblon (CRBN) ligand supplied as a hydrochloride salt for use in PROTAC synthesis and related cereblon-targeted chemical biology research. It is intended for research use and is provided with documented purity and stability data for handling and storage.
  • Thalidomide-based cereblon ligand for PROTAC synthesis.
  • Hydrochloride salt form for improved handling and solubility.
  • High purity suitable for synthetic and analytical workflows.
  • Documented stability under cold storage conditions.
  • Provided in a small-scale research package for medicinal chemistry.