Learn More
Medchemexpress LLC Thalidomide-piperazine 5-fluoride hydrochloride | 2222114-23-8 | MFCD32203462 | 98.9% | 396.80 g/mol | C17H18ClFN4O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1454365G
Encompass_Preferred
Thalidomide-piperazine 5-fluoride hydrochloride is a thalidomide-derived cereblon (CRBN) E3 ubiquitin ligase ligand supplied as the hydrochloride salt for research use. It is a light-yellow solid with high reported purity and is intended for use in PROTAC synthesis and related chemical biology applications.
  • Used as a ligand for cereblon (CRBN) in PROTAC synthesis.
  • High purity (98.9%) suitable for research applications.
  • Provided as a hydrochloride salt for improved handling.
  • Light yellow solid form for easy weighing and storage.
  • Storage: room temperature; in solvent, -80°C (2 years) or -20°C (1 year).
  • Available in multiple package sizes including gram-scale quantities.
Catalog No. 50-002-83224