Learn More
Medchemexpress LLC Thalidomide-piperazine-PEG1-COOH | 2731007-11-5 | 99.2% | 458.46 | 25 MG
SDP
Catalog No. 5000350123
Encompass_Preferred
Thalidomide-Piperazine-PEG1-COOH is a synthetic E3 ligase ligand-linker coupler containing a Thalidomide-based ligand and one linker. It acts as an E3 ligase ligand-linker conjugate.
  • E3 ligase ligand-linker coupler
  • Contains a Thalidomide-based ligand and one linker
Catalog No. 50-003-50123 Supplier Medchemexpress LLC Supplier No. 25MGHY138782
Thalidomide-Piperazine-PEG1-COOH is a synthetic E3 ligase ligand-linker coupler containing a Thalidomide-based ligand and one linker. It acts as an E3 ligase ligand-linker conjugate.
  • E3 ligase ligand-linker coupler
  • Contains a Thalidomide-based ligand and one linker