Learn More
Medchemexpress LLC Thalidomide-piperazine-PEG1-NH2 diTFA | 00-00-0 | 99.4% | C25H29F6N5O9 | 10MG
SDP
Catalog No. 5000261960
Encompass_Preferred
Thalidomide-Piperazine-PEG1-NH2 diTFA is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-based cereblon ligand with a short PEG1/piperazine linker for PROTAC design. Intended for research use only, the compound is supplied as a solid (white to yellow) with characterized solubility and storage recommendations to support experimental workflows.
  • High purity for research use (99.4%).
  • Thalidomide-based cereblon ligand facilitates E3 ligase recruitment.
  • Short PEG1/piperazine linker suitable for PROTAC assembly.
  • DMSO solubility: 100 mg/mL; sonication recommended.
  • In vivo formulation options reach ~2.5 mg/mL as suspended or clear solutions.
  • Storage: sealed at 4°C; in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-61960 Supplier Medchemexpress LLC Supplier No. HY138789A10MG
Thalidomide-Piperazine-PEG1-NH2 diTFA is a synthesized E3 ligase ligand-linker conjugate that combines a thalidomide-based cereblon ligand with a short PEG1/piperazine linker for PROTAC design. Intended for research use only, the compound is supplied as a solid (white to yellow) with characterized solubility and storage recommendations to support experimental workflows.
  • High purity for research use (99.4%).
  • Thalidomide-based cereblon ligand facilitates E3 ligase recruitment.
  • Short PEG1/piperazine linker suitable for PROTAC assembly.
  • DMSO solubility: 100 mg/mL; sonication recommended.
  • In vivo formulation options reach ~2.5 mg/mL as suspended or clear solutions.
  • Storage: sealed at 4°C; in solvent -80°C (6 months) or -20°C (1 month).