Learn More
Medchemexpress LLC Thalidomide-piperazine-PEG1-NH2 (diTFA) | 2984178-51-8 | 99.4% | 657.52 | 25 MG

Supplier: Medchemexpress LLC 25MGHY138789A
Thalidomide-Piperazine-PEG1-NH2 (diTFA) is a chemical compound suitable for laboratory research. It is stable under recommended storage conditions and is not classified as a hazardous substance.
- Molecular formula: C25H29F6N5O9
- Molecular weight: 657.52
- Purity: 99.4%
- Appearance: White to yellow solid
- Storage conditions: 4°C sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.