Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-Piperazine-PEG1-NH2 (diTFA) | 657.52 | 100 MG
SDP

Catalog No. 5000350447
Encompass_Preferred

Thalidomide-Piperazine-PEG1-NH2 (diTFA) is a chemical compound primarily intended for laboratory research and the manufacture of various substances. Presented as a solid, this product has a molecular weight of 657.52. It is designed for experienced personnel and research applications only, and has not been fully validated for medical uses. Proper storage is crucial: the recommended temperature is 4°C in sealed containers away from moisture. If stored in solvent, it can be kept at -80°C for 6 months or -20°C for 1 month, also in sealed conditions away from moisture.

  • Designed for laboratory research applications
  • Suitable for experienced personnel
  • Molecular weight of 657.52
  • Appears as a solid
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-50447 Supplier Medchemexpress LLC Supplier No. HY138789A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-Piperazine-PEG1-NH2 (diTFA) is a chemical compound primarily intended for laboratory research and the manufacture of various substances. Presented as a solid, this product has a molecular weight of 657.52. It is designed for experienced personnel and research applications only, and has not been fully validated for medical uses. Proper storage is crucial: the recommended temperature is 4°C in sealed containers away from moisture. If stored in solvent, it can be kept at -80°C for 6 months or -20°C for 1 month, also in sealed conditions away from moisture.

  • Designed for laboratory research applications
  • Suitable for experienced personnel
  • Molecular weight of 657.52
  • Appears as a solid
  • Store at 4°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.