Learn More
Medchemexpress LLC Thalidomide-piperazine-PEG2-COOH | 2797430-20-5 | 99.8% | 502.52 | 25 MG
SDP
Catalog No. 5000349216
Encompass_Preferred
Thalidomide-Piperazine-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Used as a linker in PROTAC technology
  • Related to PROTACs, an emerging therapeutic modality in precision medicine
  • Related to cereblon-based small-molecule compounds for controlling neural stem cell proliferation in regenerative medicine
Catalog No. 50-003-49216 Supplier Medchemexpress LLC Supplier No. 25MGHY138781
Thalidomide-Piperazine-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates thalidomide based cereblon ligand
  • Used as a linker in PROTAC technology
  • Related to PROTACs, an emerging therapeutic modality in precision medicine
  • Related to cereblon-based small-molecule compounds for controlling neural stem cell proliferation in regenerative medicine