Learn More
Medchemexpress LLC Thalidomide-piperazine-PEG2-COOH | 2797430-20-5 | 99.8% | C24H30N4O8 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13878110MG
Encompass_Preferred
Thalidomide-Piperazine-PEG2-COOH is a synthesized E3 ligase ligand-linker conjugate that pairs a thalidomide-derived cereblon ligand with a PEG2-containing piperazine linker. It is intended for use in proteolysis-targeting chimera (PROTAC) research and supports conjugation via a terminal carboxylic acid for downstream coupling.
  • High purity suitable for research applications.
  • Contains a carboxylic acid functional group for facile coupling.
  • Designed to recruit the cereblon E3 ligase in degrader constructs.
  • PEG2 linker provides short, flexible spacer for conjugation.
  • Characterized by molecular formula C24H30N4O8 and molecular weight 502.52 g·mol⁻¹.
Catalog No. 50-002-61627