Learn More
Medchemexpress LLC Thalidomide-Piperazine-PEG2-NH2 diTFA | 99.7% | 701.57 | 100 MG
SDP
Catalog No. 5000351730
Encompass_Preferred
Thalidomide-Piperazine-PEG2-NH2 diTFA is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a linker, which are components utilized in PROTAC technology. This product is intended for research use only. PROTACs, or Proteolysis Targeting Chimeras, function by connecting two different ligands with a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Utilized in PROTAC technology
  • For research use only
  • Functions by connecting two different ligands with a linker
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-51730 Supplier Medchemexpress LLC Supplier No. HY138788A100MG
Thalidomide-Piperazine-PEG2-NH2 diTFA is a synthesized E3 ligase ligand-linker conjugate. It incorporates a Thalidomide-based cereblon ligand and a linker, which are components utilized in PROTAC technology. This product is intended for research use only. PROTACs, or Proteolysis Targeting Chimeras, function by connecting two different ligands with a linker: one ligand binds to an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates Thalidomide-based cereblon ligand
  • Utilized in PROTAC technology
  • For research use only
  • Functions by connecting two different ligands with a linker
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins