Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-Piperazine-PEG2-NH2 (diTFA) | 99.7% | 701.57 | 50 MG
SDP

Catalog No. 5000350328
Encompass_Preferred

Thalidomide-Piperazine-PEG2-NH2 diTFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. It is for research use only.

  • Incorporates thalidomide-based cereblon ligand
  • Used in PROTAC technology to selectively degrade target proteins
  • Targets cereblon
  • Appearance: Solid, light yellow to green yellow
  • Solubility in water: 50 mg/mL (71.27 mM, requires ultrasonication)
  • Storage: 4°C sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-50328 Supplier Medchemexpress LLC Supplier No. HY138788A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-Piperazine-PEG2-NH2 diTFA is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. It is for research use only.

  • Incorporates thalidomide-based cereblon ligand
  • Used in PROTAC technology to selectively degrade target proteins
  • Targets cereblon
  • Appearance: Solid, light yellow to green yellow
  • Solubility in water: 50 mg/mL (71.27 mM, requires ultrasonication)
  • Storage: 4°C sealed, away from moisture; in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.