Learn More
Medchemexpress LLC Thalidomide-Piperazine-Piperidine hydrochloride | 2599846-44-1 | 98.4% | 461.94 | 100 MG
SDP
Catalog No. 5000352379
Encompass_Preferred
Thalidomide-Piperazine-Piperidine hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. For research use only, this product is not sold to patients.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-52379 Supplier Medchemexpress LLC Supplier No. HY138783A100MG
Thalidomide-Piperazine-Piperidine hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. For research use only, this product is not sold to patients.
  • PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein.
  • PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.