Learn More
Medchemexpress LLC Thalidomide-Piperazine-Piperidine hydrochloride | 2599846-44-1 | 98.4% | 461.94 | 50 MG
SDP
Catalog No. 5000350606
Encompass_Preferred
This product is a synthesized E3 ligase ligand-linker conjugate. It incorporates a cereblon ligand and a linker that is used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Incorporates a cereblon ligand.
  • Utilizes a linker used in PROTAC technology.
Catalog No. 50-003-50606 Supplier Medchemexpress LLC Supplier No. HY138783A50MG
Add to Cart
Add to Cart
This product is a synthesized E3 ligase ligand-linker conjugate. It incorporates a cereblon ligand and a linker that is used in PROTAC technology.
  • Synthesized E3 ligase ligand-linker conjugate.
  • Incorporates a cereblon ligand.
  • Utilizes a linker used in PROTAC technology.