Learn More
Medchemexpress LLC Thalidomide-piperazine-piperidine hydrochloride | 2599846-44-1 | 98.4% | 461.94 g/mol | C22H28ClN5O4 | 5 MG
SDP
Catalog No. 5000221444
Encompass_Preferred
Thalidomide-piperazine-piperidine hydrochloride is an E3 ligase ligand-linker conjugate derived from a thalidomide cereblon ligand and a piperazine-piperidine linker, supplied as the hydrochloride salt for use in PROTAC design and related medicinal chemistry research. It is provided as a high-purity research reagent and is not intended for clinical or human use.
  • E3 ligase ligand-linker conjugate suitable for PROTAC synthesis.
  • Supplied as the hydrochloride salt for improved handling and stability.
  • High purity, 98.4%, suitable for medicinal chemistry applications.
  • Available in small milligram pack sizes for discovery research.
  • For research use only; not for clinical or human use.
Catalog No. 50-002-21444 Supplier Medchemexpress LLC Supplier No. HY138783A5MG
Thalidomide-piperazine-piperidine hydrochloride is an E3 ligase ligand-linker conjugate derived from a thalidomide cereblon ligand and a piperazine-piperidine linker, supplied as the hydrochloride salt for use in PROTAC design and related medicinal chemistry research. It is provided as a high-purity research reagent and is not intended for clinical or human use.
  • E3 ligase ligand-linker conjugate suitable for PROTAC synthesis.
  • Supplied as the hydrochloride salt for improved handling and stability.
  • High purity, 98.4%, suitable for medicinal chemistry applications.
  • Available in small milligram pack sizes for discovery research.
  • For research use only; not for clinical or human use.