Learn More
Medchemexpress LLC Thalidomide-Piperazine-Piperidine hydrochloride | 2599846-44-1 | 98.41% | 461.94 | 25 MG
SDP
Catalog No. 5000350483
Encompass_Preferred
Thalidomide-Piperazine-Piperidine hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. This product is for research use only.
  • Incorporates thalidomide based cereblon ligand
  • Utilizes a linker used in PROTAC technology
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-50483 Supplier Medchemexpress LLC Supplier No. 25MGHY138783A
Thalidomide-Piperazine-Piperidine hydrochloride is a synthesized E3 ligase ligand-linker conjugate. It incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology. This product is for research use only.
  • Incorporates thalidomide based cereblon ligand
  • Utilizes a linker used in PROTAC technology
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins