Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thalidomide-piperazine-piperidine hydrochloride | 2599846-44-1 | MFCD34602683 | 98.4% | C22H28ClN5O4 | 10MG
SDP

Catalog No. 5000259775
Encompass_Preferred

Thalidomide-piperazine-piperidine hydrochloride is a thalidomide-derived cereblon (CRBN) E3 ligase ligand linked via a piperazine-piperidine spacer, provided as a research-use building block for targeted protein degradation and related chemical biology applications. It is supplied as a solid with high purity and defined storage conditions.

  • Thalidomide-derived cereblon E3 ligase ligand.
  • Designed as a building block for PROTAC and degrader synthesis.
  • High purity (98.4%).
  • Solid, light yellow to green yellow appearance.
  • Molecular formula C22H28ClN5O4; molecular weight 461.94.
  • Store at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-59775 Supplier Medchemexpress LLC Supplier No. HY138783A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thalidomide-piperazine-piperidine hydrochloride is a thalidomide-derived cereblon (CRBN) E3 ligase ligand linked via a piperazine-piperidine spacer, provided as a research-use building block for targeted protein degradation and related chemical biology applications. It is supplied as a solid with high purity and defined storage conditions.

  • Thalidomide-derived cereblon E3 ligase ligand.
  • Designed as a building block for PROTAC and degrader synthesis.
  • High purity (98.4%).
  • Solid, light yellow to green yellow appearance.
  • Molecular formula C22H28ClN5O4; molecular weight 461.94.
  • Store at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.