Learn More
Medchemexpress LLC Thalidomide-propargyl | 2098487-39-7 | 100.0% | 312.28 | 50 MG
SDP
Catalog No. 5000438187
Encompass_Preferred
Thalidomide-propargyl is a Thalidomide-based Cereblon ligand utilized in the recruitment of CRBN protein. It can be linked to a ligand for protein via a linker to create IMiD-containing PROTACs. This product is also a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups.
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to ligands for protein to form IMiD containing PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups
Catalog No. 50-004-38187 Supplier Medchemexpress LLC Supplier No. HY12645750MG
Thalidomide-propargyl is a Thalidomide-based Cereblon ligand utilized in the recruitment of CRBN protein. It can be linked to a ligand for protein via a linker to create IMiD-containing PROTACs. This product is also a click chemistry reagent, featuring an Alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups.
  • Thalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to ligands for protein to form IMiD containing PROTACs
  • Click chemistry reagent
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups